missing translation for 'onlineSavingsMsg'
Learn More

Ammonium bismuth citrate, Bi 48-52%, water ca 2%

Product Code. 11488846
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488846 250 g 250g
11478846 50 g 50g
11468846 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488846 Supplier Thermo Scientific Alfa Aesar Supplier No. A17003.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25530-63-6 | C6H11BiNO7 | 418.134 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25530-63-6
Molecular Formula C6H11BiNO7
Molecular Weight (g/mol) 418.134
MDL Number MFCD00036420
InChI Key JDINTLWFPXWXCT-UHFFFAOYSA-N
Synonym ammonium bismuth citrate
PubChem CID 87101692
IUPAC Name azane;bismuth;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Bi]

Specifications

Quantity 250 g
Formula Weight 433.12
Assay Bi 48 to 52%, water ≈2%
Chemical Name or Material Ammonium bismuth citrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.