missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Bicarbonate, 0.1 M, Ricca Chemical

Product Code. 31751097
Change view
Click to view available options
Quantity:
4 L
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31751097 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31751097 Supplier Ricca Chemical Company Supplier No. R06216504A
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Colorless
CAS 1066-33-7, 7732-18-5
CAS Min % 97.0, 0.77
CAS Max % 100.0, 0.8
Packaging Natural Poly Bottle
Molecular Formula CH5NO3
pH 7.8
MDL Number MFCD00012138
Quantity 4 L
Physical Form Liquid
Concentration or Composition Notes 0.1Molar
Synonym ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
InChI Key ATRRKUHOCOJYRX-UHFFFAOYSA-N
SMILES N.OC(O)=O
IUPAC Name carbonic acid amine
Molecular Weight (g/mol) 79.06
PubChem CID 14013
Grade Laboratory
Chemical Name or Material Ammonium Bicarbonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.