missing translation for 'onlineSavingsMsg'
Learn More

Ammonium benzoate, 99%

Product Code. 11307348
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
This item is not returnable. View return policy

Product Code. 11307348

Brand: Thermo Scientific Alfa Aesar 030753.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1863-63-4 | C7H9NO2 | 139.154 g/mol

Ammonium benzoate is used predominantly in industries which acts as a corrosion inhibitors and anti-scaling agents . It is also used as paint additives and coating additives. Other consumer uses include cleaning and furnishing Care products, paints and coatings, personal care products, water treatment products. Ammonium benzoate is also used as chemical analysis reagents, in electronics industry, used for the production of electrolytic capacitors and also used in pharmaceutical industries. Used as adhesives, metal rust treatment and rubber additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1863-63-4
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00013069
InChI Key VWSRWGFGAAKTQG-UHFFFAOYSA-N
Synonym ammonium benzoate, benzoic acid, ammonium salt, ammonii benzoas, ammonium benzoicum, vulnoc ab, benzoic acid ammonium salt, hsdb 555, unii-ac80wd7gpf, benzoic acid, ammonium salt 1:1, ac80wd7gpf
PubChem CID 15830
IUPAC Name azanium;benzoate
SMILES C1=CC=C(C=C1)C(=O)[O-].[NH4+]

Specifications

Melting Point 192°C to 198°C (decomposition)
Color White
Physical Form Crystalline
Quantity 500 g
Flash Point 110°C (230°F)
Merck Index 14,496
Solubility Information It is soluble in water,methanol insoluble in diethyl ether.
Formula Weight 139.15
Percent Purity 99%
Odor Weak
Sensitivity Hygroscopic
Chemical Name or Material Ammonium benzoate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.