missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Aluminum Sulfate Dodecahydrate, purum p.a., ≥99.0% (T), Honeywell™

Product Code. 15670880 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15670880 1 kg Poly bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15670880 Supplier Honeywell Research Chemicals Supplier No. 096941KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥99.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 93.5°C
CAS 7784-26-1
Color White
pH 2.6 (at 100g/L, 25°C)
Physical Form Crystalline
Quantity 1 kg
Molecular Formula AlH28NO20S2
Linear Formula AlNH4(SO4)2 · 12H2O
MDL Number MFCD00149958
UN Number NONH for all modes of transport
Synonym ammonia alum, unii-5c36drl9zn, aluminum ammonium sulfate dodecahydrate, aluminum ammonium disulfate dodecahydrate, ammonium aluminum sulfate hydrate, alum, ammonium usp, aluminum ammonium sulfate dodecahydrate, ammonium aluminum sulfate dodecahydrate, 5c36drl9zn, ammonium aluminum disulfate dodecahydrate
InChI Key WZUKKIPWIPZMAS-UHFFFAOYSA-K
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3]
IUPAC Name aluminum;azanium;disulfate;dodecahydrate
Molecular Weight (g/mol) 453.313
PubChem CID 62668
Formula Weight 453.33g/mol
Percent Purity ≥99.0% (T)
Grade purum p.a.
Odor Odorless
Packaging Poly bottle
Density ∽1.640g/cu.cm at 20°C
Chemical Name or Material Ammonium Aluminum Sulfate Dodecahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.