missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, HPLC Grade, J.T. Baker™

Product Code. 15513351 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15513351 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15513351 Supplier Avantor J.T.Baker Supplier No. 059908
  • ONLINE EXCLUSIVE PRICE
    £552.00
    £444.63 / 1kg
    Save £107.37
    19% Off
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High purity solvents for use in analytical and preparative separations

    For Use in High Performance Liquid Chromatography; For Laboratory,Research,or Manufacturing Use

Specifications

CAS 631-61-8
Quantity 1 kg
Molecular Formula C2H7NO2
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES CC(=O)[O-].[NH4+]
IUPAC Name azanium;acetate
Molecular Weight (g/mol) 77.083
PubChem CID 517165
ChEBI CHEBI:62947
Formula Weight 77.08
Grade HPLC
Packaging Poly Bottle
Specific Gravity 1L = 1.07kg
Chemical Name or Material Ammonium Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.