missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, Extra Pure, SLR, Crystals, Fisher Chemical

Product Code. 10050030 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10050030 1 kg Plastic powder jar 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10050030 Supplier Fisher Chemical Supplier No. A/3400/60

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 631-61-8
Melting Point 114°C
Color White
pH 7
Packaging Plastic powder jar
Molecular Formula C2H7NO2
MDL Number 13066
Quantity 1 kg
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES CC(=O)[O-].[NH4+]
IUPAC Name azanium;acetate
Molecular Weight (g/mol) 77.083
PubChem CID 517165
ChEBI CHEBI:62947
Formula Weight 77.08g/mol
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.