missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10518610 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
500 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
250g
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10518610 5 kg Plastic keg 5kg
10203900 250 g Plastic powder jar 250g
10365260 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10518610 Supplier Fisher Chemical Supplier No. A/3440/65
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

C2H7NO2, CAS Number-631-61-8, 500g, White, 77.08g/mol, 114 deg.C, 13066, Plastic powder jar, Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
MDL Number 13066

Specifications

Melting Point 114°C
Color White
pH 7
Packaging Plastic keg
Quantity 5 kg
Formula Weight 77.08g/mol
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.