missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate Anhydrous, ACS Reagent Grade, MP Biomedicals™

Product Code. 11480482 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kg
Unit Size:
1kg

Product Code. 11480482

Brand: MP Biomedicals 0219140491

Please to purchase this item. Need a web account? Register with us today!

Specifications

CAS 631-61-8
Quantity 1 kg
Molecular Formula C2H7NO2
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES CC(=O)[O-].[NH4+]
IUPAC Name azanium;acetate
Molecular Weight (g/mol) 77.083
PubChem CID 517165
ChEBI CHEBI:62947
Formula Weight 77.1
Grade ACS Reagent
Density 1.17g/cm3
Chemical Name or Material Ammonium Acetate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.