missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Ammonium Acetate, Ultra Pure Grade

Product Code. 17128548 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 17128548

Brand: MP Biomedicals™ 0219384880

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ammonium Acetate, Ultra Pure Grade is used in the purification and precipitation of DNA and for protein crystallization.

  • Ammonium acetate, ultra pure, is for applications which require tight control of elemental content.
  • Ammonium acetate is a widely used reagent in molecular biology and chromatography.
  • Its applications include the purification and precipitation of DNA and protein crystallization.
  • Ammonium acetate is commonly used in HPLC and MS analysis of various compounds, such as oligosaccharides, proteins, and peptides.
  • Ammonium acetate is also used for the nonaqueous capillary electrophoresis-mass spectrometry (NACE-MS) of lipophilic peptides and therapeutic drugs.
TRUSTED_SUSTAINABILITY

Specifications

Assay ≥97%
Chemical Name or Material Ammonium Acetate
CAS 631-61-8
Content And Storage Room Temperature
Density 1.17 at 68° F
Formula Weight 77.1
Melting Point 237.2° F
Molecular Formula C2H7NO2
Purity Ultrapure
Quantity 100 g
Synonym unii-rre756s6q2
Form Crystalline Powder
Show More Show Less

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.