missing translation for 'onlineSavingsMsg'
Learn More

Ammonium 7-Fluoro-2,1,3-benzoxadiazole-4-sulfonate, TRC

Product Code. 30333257
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30333257 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333257 Supplier Toronto Research Chemicals Supplier No. TRCA6340001MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium 7-Fluoro-2,1,3-benzoxadiazole-4-sulfonate
CAS 84806-27-9
Molecular Formula C6 H3 F N2 O4 S . H3 N
Quantity 1 mg
InChI Formula InChI=1S/C6H3FN2O4S.H3N/c7-3-1-2-4(14(10,11)12)6-5(3)8-13-9-6;/h1-2H,(H,10,11,12);1H3
SMILES N.OS(=O)(=O)c1ccc(F)c2nonc12
IUPAC Name azane;7-fluoro-2,1,3-benzoxadiazole-4-sulfonic acid
Molecular Weight (g/mol) 235.19
Formula Weight 235.0063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.