missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Solution, 35%, Certified AR for Analysis, d=0.88, Fisher Chemical™

Catalog Number 10305220
27.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
HDPE plastic bottle
Plastic Coated Glass bottle
Plastic container
1336-21-6
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
14923
CHEBI:18219
N.O
1336-21-6
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
14923
CHEBI:18219
N.O

H5NO, CAS Number-1336-21-6, aquammonia, ammonium hydroxide solution, ammonium hydroxide nh4 oh, ammonia water, aqua ammonia, ammonia, monohydrate, ammonium hydroxide, ammonia aqueous, household ammonia, ammonia, aqua, 2.5L, CHEBI:18219, Colorless, 35.05g/mol, VHUUQVKOLVNVRT-UHFFFAOYSA-N

TRUSTED_SUSTAINABILITY
pH 12
Color Colorless
Quantity 1 L
IUPAC Name azanium;hydroxide
Formula Weight 35.05g/mol
Packaging Plastic Coated Glass bottle
Physical Form Liquid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'