missing translation for 'onlineSavingsMsg'
Learn More

Ammonia, Dilute R1, EP Grade, Reagecon™
BIOPHARMA

Product Code. 12603623 Shop All Reagecon Products
Click to view available options
Quantity:
1 L
Packaging:
Tamper evident
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 12603623

Brand: Reagecon 1004702

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured according to European Pharmacopoeia Chapter 4 (4.1.1) and prepared in an ISO 9001 facility. Ammonia, Dilute R1, EP Grade, Reagecon™ is a ready-to-use analytical volumetric solution for use in manual or automatic titrations. It contains between 100–104g/L of NH3.

  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.1.1) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use reagent eliminates the time and labor of preparing precise volumetric solutions

Specifications

CAS 1336-21-6, 7732-18-5
Physical Form Liquid
Quantity 1 L
Molecular Formula H5NO
MDL Number MFCD00066650
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O
IUPAC Name azanium;hydroxide
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Formula Weight 35.05g/mol
Grade Eu. Ph.
Concentration or Composition (by Analyte or Components) 100–104g/L NH3
Packaging Tamper evident
Chemical Name or Material Ammonia Hydroxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • AMMONIA DILUTE R1
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : ammonia
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.