missing translation for 'onlineSavingsMsg'
Learn More

Ammeline, TRC

Product Code. 30338845
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30338845 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338845 Supplier Toronto Research Chemicals Supplier No. TRCA63366050MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,6-Diamino-2-hydroxy-1,3,5-triazine
CAS 645-92-1
Molecular Formula C3 H5 N5 O
Quantity 50 mg
Synonym Ammeline (6CI), s-Triazin-2(1H)-one, 4,6-diamino- (7CI), s-Triazin-2-ol, 4,6-diamino- (8CI), 4,6-Diamino-1,3,5-triazin-2(1H)-one, 2,4-Diamino-1,3,5-triazin-6-one, 2,4-Diamino-6-hydroxy-1,3,5-triazine, 2,4-Diamino-6-hydroxy-s-triazine, 2-Hydroxy-4,6-diamino-s-triazine, 2-Hydroxydesethyldesisopropylatrazine, 4,6-Diamino-2-hydroxy-s-triazine, Ammelin, Deethyldeisopropylhydroxyyatrazine, GS 11791, NSC 9778, s-Diaminohydroxytriazine
InChI Formula InChI=1S/C3H5N5O/c4-1-6-2(5)8-3(9)7-1/h(H5,4,5,6,7,8,9)
SMILES Nc1nc(N)nc(O)n1
IUPAC Name 4,6-diamino-1,3,5-triazin-2-ol
Molecular Weight (g/mol) 127.1
Formula Weight 127.0494
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.