missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMMECR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530555
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, Raji. Predicted reactivity: Mouse (100%), Zebrafish (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The exact function of this gene is not known, however, submicroscopic deletion of the X chromosome including this gene, COL4A5, and FACL4 genes, result in a contiguous gene deletion syndrome, the AMME complex (Alport syndrome, mental retardation, midface hypoplasia, and elliptocytosis). Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| AMMECR1 | |
| Polyclonal | |
| Unconjugated | |
| AMMECR1 | |
| 6230420G18Rik; Alport syndrome mental retardation midface hypoplasia and elliptocytosis chromosomal region protein 1; Alport syndrome, mental retardation, midface hypoplasia and elliptocytosis chromosomal region gene 1; Alport syndrome, mental retardation, midface hypoplasia and elliptocytosis chromosomal region gene 1 homolog; AMME syndrome candidate gene 1 protein; AMME syndrome candidate gene 1 protein homolog; Ammecr1; AMMERC1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9949 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9Y4X0 | |
| AMMECR1 | |
| Recombinant protein encompassing a sequence within the center region of human AMMECR1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction