missing translation for 'onlineSavingsMsg'
Learn More

Amlodipine, TRC

Product Code. 18074010
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18074010 1 g 1g
18043920 100 mg 100mg
17919829 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18074010 Supplier Toronto Research Chemicals Supplier No. TRCA6334951G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88150-42-9
Molecular Formula C20 H25 Cl N2 O5
Molecular Weight (g/mol) 408.88
Synonym 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5-Pyridinedicarboxylic acid 3-ethyl 5-methyl ester, (R,S)-Amlodipine, 2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-3-(ethoxycarbonyl)-5-(methoxycarbonyl)-6-methyl-1,4-dihydropyridine, Amlodipine, Pelmec, Racemic Amlodipine
IUPAC Name 3-O-ethyl 5-O-methyl 2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(COCCN)NC(=C(C1c2ccccc2Cl)C(=O)OC)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H25ClN2O5/c1-4-28-20(25)18-15(11-27-10-9-22)23-12(2)16(19(24)26-3)17(18)13-7-5-6-8-14(13)21/h5-8,17,23H,4,9-11,22H2,1-3H3
Formula Weight 408.1452
Percent Purity >95%
Chemical Name or Material Amlodipine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.