missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amlodipine, MedChemExpress
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
Amlodipine, an antianginal agent and an orally active dihydropyridine calcium channel blocker, works by blocking the voltage-dependent L-type calcium channels, thereby inhibiting the initial influx of calcium. Amlodipine can be used for the research of high blood pressure and cancer.
Specifications
Specifications
| Chemical Name or Material | Amlodipine |
| CAS | 88150-42-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H25ClN2O5 |
| Quantity | 1 g |
| Solubility Information | DMSO : 31.25 mg/mL (76.43 mM; Need ultrasonic) |
| SMILES | O=C(C1=C(COCCN)NC(C)=C(C(OC)=O)C1C2=CC=CC=C2Cl)OCC |
| Molecular Weight (g/mol) | 408.88 |
| Formula Weight | 408.88 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction