missing translation for 'onlineSavingsMsg'
Learn More

Amlodipine Maleate, TRC

Product Code. 30345656
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30345656

Brand: Toronto Research Chemicals TRCA633595100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amlodipine Maleate
CAS 88150-47-4
Purity Grade Notes HPLC
Molecular Formula C20 H25 Cl N2 O5 . C4 H4 O4
Quantity 100 mg
Synonym Amlodipine Maleate, 3,5-Pyridinedicarboxylic acid, 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-, 3-ethyl 5-methyl ester, (2Z)-2-butenedioate (1:1), 2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5-pyridinedicarboxylic acid 3-ethyl 5-methyl ester (Z)-2-butenedioate (1:1), Amlodipine maleate, UK 48340-11
InChI Formula InChI=1S/C20H25ClN2O5.C4H4O4/c1-4-28-20(25)18-15(11-27-10-9-22)23-12(2)16(19(24)26-3)17(18)13-7-5-6-8-14(13)21;5-3(6)1-2-4(7)8/h5-8,17,23H,4,9-11,22H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
SMILES CCOC(=O)C1=C(COCCN)NC(=C(C1c2ccccc2Cl)C(=O)OC)C.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;3-O-ethyl 5-O-methyl 2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Weight (g/mol) 524.95
Formula Weight 524.1562
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.