missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amlodipine Maleate, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Amlodipine Maleate
Specifications
Specifications
| Chemical Name or Material | Amlodipine Maleate |
| CAS | 88150-47-4 |
| Analyte or Component Names | Amlodipine Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H25 Cl N2 O5 . C4 H4 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Amlodipine Maleate, 2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5-pyridinedicarboxylic acid 3-ethyl 5-methyl ester (Z)-2-butenedioate (1:1), Amlodipine maleate, UK 48340-11 |
| InChI Formula | InChI=1S/C20H25ClN2O5.C4H4O4/c1-4-28-20(25)18-15(11-27-10-9-22)23-12(2)16(19(24)26-3)17(18)13-7-5-6-8-14(13)21;5-3(6)1-2-4(7)8/h5-8,17,23H,4,9-11,22H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | CCOC(=O)C1=C(COCCN)NC(=C(C1c2ccccc2Cl)C(=O)OC)C.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;3-O-ethyl 5-O-methyl 2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction