missing translation for 'onlineSavingsMsg'
Learn More

Amlodipine besylate, MedChemExpress

Product Code. p-200099141
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16420878

Brand: Medchem Express HYB0317B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Amlodipine besylate (Amlodipine benzenesulfonate), an antianginal agent and an orally active dihydropyridine calcium channel blocker, works by blocking the voltage-dependent L-type calcium channels, thereby inhibiting the initial influx of calcium. Amlodipine besylate can be used for the research of high blood pressure and cancer.

Amlodipine besylate (Amlodipine benzenesulfonate), an antianginal agent and an orally active dihydropyridine calcium channel blocker, works by blocking the voltage-dependent L-type calcium channels, thereby inhibiting the initial influx of calcium. Amlodipine besylate can be used for the research of high blood pressure and cancer.

Specifications

Chemical Name or Material Amlodipine besylate
CAS 111470-99-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H31ClN2O8S
Quantity 5 g
Synonym Amlodipine benzenesulfonate
Solubility Information DMSO : ≥ 45 mg/mL (79.36 mM) ∣H2O : 1 mg/mL (1.76 mM; Need ultrasonic)
SMILES O=C(C1=C(COCCN)NC(C)=C(C(OC)=O)C1C2=CC=CC=C2Cl)OCC.O=S(C3=CC=CC=C3)(O)=O
Molecular Weight (g/mol) 567.05
Formula Weight 567.05
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.