missing translation for 'onlineSavingsMsg'
Learn More

Amlodipine besylate, MedChemExpress

Product Code. 16420878
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16420878 5 g 5g
16412139 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16420878 Supplier Medchem Express Supplier No. HYB0317B
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Amlodipine besylate (Amlodipine benzenesulfonate), an antianginal agent and an orally active dihydropyridine calcium channel blocker, works by blocking the voltage-dependent L-type calcium channels, thereby inhibiting the initial influx of calcium. Amlodipine besylate can be used for the research of high blood pressure and cancer.

Amlodipine besylate (Amlodipine benzenesulfonate), an antianginal agent and an orally active dihydropyridine calcium channel blocker, works by blocking the voltage-dependent L-type calcium channels, thereby inhibiting the initial influx of calcium. Amlodipine besylate can be used for the research of high blood pressure and cancer.

Specifications

Chemical Name or Material Amlodipine besylate
CAS 111470-99-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H31ClN2O8S
Quantity 5 g
Synonym Amlodipine benzenesulfonate
Solubility Information DMSO : ≥ 45 mg/mL (79.36 mM) ∣H2O : 1 mg/mL (1.76 mM; Need ultrasonic)
SMILES O=C(C1=C(COCCN)NC(C)=C(C(OC)=O)C1C2=CC=CC=C2Cl)OCC.O=S(C3=CC=CC=C3)(O)=O
Molecular Weight (g/mol) 567.05
Formula Weight 567.05
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.