missing translation for 'onlineSavingsMsg'
Learn More

Amlodipine besylate, 98+%, Thermo Scientific™

An L-type calcium channel antagonist with potent antioxidant activity

Brand:  Thermo Scientific Alfa Aesar J62164.03


Quantity 1g

 View more versions of this product

Product Code. 15454589

  • £75.30 / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Amlodipine besylate is used as a dihydropyridine calcium channel blocker. It is also used as an antianginal and antihypertensive. Amlodipine besylate also displays vasoprotective effects in cardiovascular disease and inhibits Ca2+-induced contractions in depolarized rat aorta (IC50 = 1.9 nM). Amlodipine besylate has been shown to inhibit proliferation of human vascular smooth muscle cells and epidermoid carcinoma A431 cells.

Solubility
Soluble in DMSO (≥20 mg/mL), water (10 mM), chloroform, ethanol (≥14 mg/mL), and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

111470-99-6
567.05
ZPBWCRDSRKPIDG-UHFFFAOYSA-N
60496
benzenesulfonic acid;3-O-ethyl 5-O-methyl 2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
C26H31ClN2O8S
MFCD00887594
amlodipine besylate, amlodipine besilate, cardiorex, amlodipine benzenesulfonate, istin, amdepin, amdipin, amlodin, amlogard, amlopin
CHEBI:2669
CCOC(=O)C1=C(NC(=C(C1C2=CC=CC=C2Cl)C(=O)OC)C)COCCN.C1=CC=C(C=C1)S(=O)(=O)O

Specifications

111470-99-6
MFCD00887594
14,491
Soluble in DMSO (?20mg/mL),water (10 mM),chloroform,ethanol (?14mg/mL),and methanol.
CCOC(=O)C1=C(NC(=C(C1C2=CC=CC=C2Cl)C(=O)OC)C)COCCN.C1=CC=C(C=C1)S(=O)(=O)O
567.05
CHEBI:2669
≥98%
C26H31ClN2O8S
1g
amlodipine besylate, amlodipine besilate, cardiorex, amlodipine benzenesulfonate, istin, amdepin, amdipin, amlodin, amlogard, amlopin
ZPBWCRDSRKPIDG-UHFFFAOYSA-N
benzenesulfonic acid;3-O-ethyl 5-O-methyl 2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
60496
567.05
Amlodipine besylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Amlodipine besylate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contain: Amlodipine besylate

SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.