missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amitriptyline Hydrochloride, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Amitriptyline Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Amitriptyline Hydrochloride |
| CAS | 549-18-8 |
| Analyte or Component Names | Amitriptyline Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H23 N . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Amitriptyline Hydrochloride, Nortriptyline Hydrochloride Imp. F (EP) as Hydrochloride, 1-Propanamine, 3-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-dimethyl-, hydrochloride (1:1), 1-Propanamine, 3-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-dimethyl-, hydrochloride (9CI), 5-(3-Dimethylaminopropylidene)dibenzo[a,d][1,4]cycloheptadiene hydrochloride, ADT, Adepril, Amicen, Amil 25, Amilent, Amilit, Amineurin, Amiplin, Amiprin, Amitid, Amitril, Amitrip, Amitriptyline chloride, Amyline, Amyzol, Anapsique, Apo-Amitriptyline, Damilen hydrochloride, Deprex, Domical, Elatrol, Elatrolet, Elavil, Enafon, Endep, Etravil, Euplit, Lantron, Laroxyl, Larozyl, Lentizol, Miketorin, NIH 10794, Noriline, Novoprotect, Pinsanu, Pinsaun, Redomex, Ro 4-1575, Saroten, Saroten Retard, Sarotena, Sarotex, Sylvemid, Syneudon, Teperin, Trepiline, Tridep, Tripta, Triptizol, Trynol, Tryptal, Tryptanol, Tryptine, Tryptizol, Trytomer, Uxen, Vanatrip, 5H-Dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine, 10,11-dihydro-N,N-dimethyl-, hydrochloride (6CI,8CI), 10,11-Dihydro-N,N-dimethyl-5H-dibenzo[a,d]cycloheptene-Delta5,gamma-propylamine hydrochloride, Nortriptyline Hydrochloride Impurity F as Hydrochloride, Nortriptyline Impurity F as Hydrochloride |
| InChI Formula | InChI=1S/C20H23N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-12H,7,13-15H2,1-2H3;1H |
| SMILES | Cl.CN(C)CCC=C1c2ccccc2CCc3ccccc13 |
| IUPAC Name | 3-(5,6-dihydrodibenzo[2,1-b:1',2'-e][7]annulen-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction