missing translation for 'onlineSavingsMsg'
Learn More

Amitriptyline EP Impurity G (Nortriptyline EP Impurity I), Mikromol™

Product Code. 30050446
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050446 25 mg 25mg
30051443 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050446 Supplier Mikromol Supplier No. MM0062.140025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Amitriptyline EP Impurity G (Nortriptyline EP Impurity I)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10,11-Dihydro-5H-dibenzo[a,d][7]-annulen-5-ol (Dibenzosuberol)
CAS 1210-34-0
Analyte or Component Names 10,11-Dihydro-5H-dibenzo[a,d][7]-annulen-5-ol (Dibenzosuberol)
Shipping Condition Room Temperature
Molecular Formula C15 H14 O
Certifications/Compliance ISO 9001
Synonym 10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-ol (Dibenzosuberol), Amitriptyline Hydrochloride Imp. G (EP), Amitriptyline Imp. G (EP), 10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-ol, 10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-ol, 5-Hydroxy-10,11-dihydro-5H-dibenzo[a,d]cycloheptene, Dibenzosuberol, NSC 86157, Amitriptyline Hydrochloride Impurity G, Nortriptyline Hydrochloride Impurity I, Amitriptyline Impurity G, Nortriptyline Impurity I
InChI Formula InChI=1S/C15H14O/c16-15-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)15/h1-8,15-16H,9-10H2
SMILES OC1c2ccccc2CCc3ccccc13
IUPAC Name 6,11-dihydro-5H-dibenzo[2,1-b:1',2'-e][7]annulen-11-ol
Molecular Weight (g/mol) 210.27
Quantity 25 mg
Formula Weight 210.1045 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.