missing translation for 'onlineSavingsMsg'
Learn More

Amitriptyline EP Impurity D (Amitriptyline USP Related Compound B), Mikromol™

Product Code. 30051525
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051525 25 mg 25mg
17951826 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051525 Supplier Mikromol Supplier No. MM0062.030025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Amitriptyline EP Impurity D (Amitriptyline USP Related Compound B)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-[3-(Dimethylamino)propyl]-10,11-dihydro-5H-dibenzo[a,d][7]annulen-5-ol
CAS 1159-03-1
Analyte or Component Names 5-[3-(Dimethylamino)propyl]-10,11-dihydro-5H-dibenzo[a,d][7]annulen-5-ol
Shipping Condition Room Temperature
Molecular Formula C20 H25 N O
Certifications/Compliance ISO 17025
Synonym Amitriptyline Hydrochloride Imp. D (EP), Amitriptyline Imp. D (EP), Nortriptyline Hydrochloride Imp. D (EP), Nortriptyline Imp. D (EP), Amitriptyline USP Related Compound B, Amitriptyline USP RC B, 5-[3-(Dimethylamino)propyl]-10,11-dihydro-5H-dibenzo[a,d][7]annulen-5-ol, Amitriptyline Related Compound B, Amitriptyline Hydrochloride Impurity D, Nortriptyline Hydrochloride Impurity D, Amitriptyline Impurity D, Nortriptyline Impurity D
InChI Formula InChI=1S/C20H25NO/c1-21(2)15-7-14-20(22)18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)20/h3-6,8-11,22H,7,12-15H2,1-2H3
SMILES CN(C)CCCC1(O)c2ccccc2CCc3ccccc13
IUPAC Name 11-[3-(dimethylamino)propyl]-5,6-dihydrodibenzo[2,1-b:1',2'-e][7]annulen-11-ol
Molecular Weight (g/mol) 295.42
Quantity 25 mg
Formula Weight 295.1936 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.