missing translation for 'onlineSavingsMsg'
Learn More

Amitifadine hydrochloride, MedChemExpress

Product Code. 16429987
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16429987

Brand: Medchem Express HY18332A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Amitifadine hydrochloride is a serotonin-norepinephrine-dopamine reuptake inhibitor (SNDRI), with IC50s of 12, 23, 96 nM for serotonin, norepinephrine and dopamine in HEK 293 cells , respectively.

Amitifadine hydrochloride is a serotonin-norepinephrine-dopamine reuptake inhibitor (SNDRI), with IC50s of 12, 23, 96 nM for serotonin, norepinephrine and dopamine in HEK 293 cells , respectively.

Specifications

Chemical Name or Material Amitifadine hydrochloride
CAS 410074-74-7
Color White
Purity Grade Notes Research
Molecular Formula C11H12Cl3N
Quantity 5 mg
Synonym DOV-21947 hydrochloride EB-1010 hydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (377.96 mM) ∣H2O : 50 mg/mL (188.98 mM; Need ultrasonic)
SMILES ClC(C=C1[C@]2(CNC3)[C@@H]3C2)=C(C=C1)Cl.[H]Cl
Molecular Weight (g/mol) 264.58
Formula Weight 264.58
Percent Purity 99.86%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.