missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amitifadine hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Amitifadine hydrochloride is a serotonin-norepinephrine-dopamine reuptake inhibitor (SNDRI), with IC50s of 12, 23, 96 nM for serotonin, norepinephrine and dopamine in HEK 293 cells , respectively.
Specifications
Specifications
| Chemical Name or Material | Amitifadine hydrochloride |
| CAS | 410074-74-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C11H12Cl3N |
| Quantity | 50 mg |
| Synonym | DOV-21947 hydrochloride EB-1010 hydrochloride |
| Solubility Information | DMSO : ≥ 100 mg/mL (377.96 mM) ∣H2O : 50 mg/mL (188.98 mM; Need ultrasonic) |
| SMILES | ClC(C=C1[C@]2(CNC3)[C@@H]3C2)=C(C=C1)Cl.[H]Cl |
| Molecular Weight (g/mol) | 264.58 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction