missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Amisyn Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570063
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
STXBP6 binds components of the SNARE complex.
Specifications
| Amisyn | |
| Polyclonal | |
| Unconjugated | |
| Stxbp6 | |
| Amisyn; BC024598; C85317; FLJ39638; HSPC156; OTTHUMP00000178754; stxbp6; syntaxin binding protein 6; syntaxin binding protein 6 (amisyn); syntaxin-binding protein 6; Unknown (protein for MGC:134570); wu:fd20e06; zgc:86678 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 217517 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8R3T5 | |
| Stxbp6 | |
| synthetic peptide directed towards the middle region of human Amisyn (aa 5-54). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction