missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Amisyn Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551960
This item is not returnable.
View return policy
Description
Immunogen sequence: NKKPTQASIT KVKQFEGSTS FVRRSQWMLE QLRQVNGIDP NGDSAEFDLL FENAFDQWVA STASEKCTFF QILHHTCQRY LTDRKPEFIN CQSKIMGGNS ILHSAADSVT SAVQKASQAL NERGERLGRA EEKTEDLK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
STXBP6 binds components of the SNARE complex.
Specifications
| Amisyn | |
| Polyclonal | |
| Unconjugated | |
| Stxbp6 | |
| Amisyn; BC024598; C85317; FLJ39638; HSPC156; OTTHUMP00000178754; stxbp6; syntaxin binding protein 6; syntaxin binding protein 6 (amisyn); syntaxin-binding protein 6; Unknown (protein for MGC:134570); wu:fd20e06; zgc:86678 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29091 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NFX7 | |
| Stxbp6 | |
| Recombinant protein corresponding to Human Amisyn. Recombinant protein control fragment (Product #RP-89139). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction