missing translation for 'onlineSavingsMsg'
Learn More

Amiodarone hydrochloride, Thermo Scientific™

Product Code. 15592223 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15592223

Brand: Thermo Scientific Acros 457980010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 19774-82-4
Molecular Formula C25H29I2NO3·HCl
Molecular Weight (g/mol) 681.77
InChI Key ITPDYQOUSLNIHG-UHFFFAOYSA-N
Synonym amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron
PubChem CID 441325
IUPAC Name (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride
SMILES CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl

Specifications

Melting Point 156.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 681.77
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Amiodarone hydrochloride
compliance-icons compliance-icons
Product Identifier
  • Amiodarone hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 2-Butyl-3-benzofuryl4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl ketone hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.