missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amiodarone hydrochloride, MedChemExpress
Description
Amiodarone hydrochloride, a benzofuran-based Class III antiarrhythmic agent, inhibits WT outwardIhERG tails with an IC50 of ∼45 nM. Amiodarone hydrochloride induces cell proliferation and myofibroblast differentiation via ERK1/2 and p38 MAPK signaling in fibroblasts. Amiodarone hydrochloride can be used in the research of both supraventricular and ventricular arrhythmias.
Specifications
Specifications
| Chemical Name or Material | Amiodarone hydrochloride |
| CAS | 19774-82-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H30ClI2NO3 |
| Quantity | 1 g |
| Solubility Information | DMSO : 24.5 mg/mL (35.94 mM; Need ultrasonic and warming) |
| SMILES | CCCCC1=C(C(C2=CC(I)=C(OCCN(CC)CC)C(I)=C2)=O)C3=C(O1)C=CC=C3.Cl |
| Molecular Weight (g/mol) | 681.77 |
| Formula Weight | 681.77 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?