missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amiodarone Hydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Amiodarone Hydrochloride |
| CAS | 19774-82-4 |
| Analyte or Component Names | Amiodarone Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H29 I2 N O3 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Amiodarone Hydrochloride, (2-Butyl-3-benzofuranyl)[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone hydrochloride, 2-Butyl-3-benzofuranyl 4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl ketone hydrochloride, Amiodar, Amiodarone hydrochloride, Amiodaronum hydrochloride, Ancoron, Angiodarona, Atlansil, Cordarex, Cordarone, Cordarone X, L 3428, L 3428-Labaz, Miocard, Miodaron, Ortacrone, Pacerone, Ritmocardyl, Rythmarone, SKF 33134A, Trangorex |
| InChI Formula | InChI=1S/C25H29I2NO3.ClH/c1-4-7-11-22-23(18-10-8-9-12-21(18)31-22)24(29)17-15-19(26)25(20(27)16-17)30-14-13-28(5-2)6-3;/h8-10,12,15-16H,4-7,11,13-14H2,1-3H3;1H |
| Solution Type | Neat |
| SMILES | Cl.CCCCc1oc2ccccc2c1C(=O)c3cc(I)c(OCCN(CC)CC)c(I)c3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?