missing translation for 'onlineSavingsMsg'
Learn More

Amiodarone Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30459480
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459480 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459480 Supplier LoGiCal Supplier No. LGCAMP0697.0001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amiodarone Hydrochloride
CAS 19774-82-4
Analyte or Component Names Amiodarone Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C25 H29 I2 N O3 . Cl H
Certifications/Compliance ISO 17034
Synonym Amiodarone Hydrochloride, (2-Butyl-3-benzofuranyl)[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone hydrochloride, 2-Butyl-3-benzofuranyl 4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl ketone hydrochloride, Amiodar, Amiodarone hydrochloride, Amiodaronum hydrochloride, Ancoron, Angiodarona, Atlansil, Cordarex, Cordarone, Cordarone X, L 3428, L 3428-Labaz, Miocard, Miodaron, Ortacrone, Pacerone, Ritmocardyl, Rythmarone, SKF 33134A, Trangorex
InChI Formula InChI=1S/C25H29I2NO3.ClH/c1-4-7-11-22-23(18-10-8-9-12-21(18)31-22)24(29)17-15-19(26)25(20(27)16-17)30-14-13-28(5-2)6-3;/h8-10,12,15-16H,4-7,11,13-14H2,1-3H3;1H
Solution Type Single Solution
SMILES Cl.CCCCc1oc2ccccc2c1C(=O)c3cc(I)c(OCCN(CC)CC)c(I)c3
IUPAC Name (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 681.77
Quantity 1 mL
Formula Weight 681.0004 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.