missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Amintra (R) Cobalt NTA Resin

Product Code. p-7180204 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
100 mL
Unit Size:
100mL
10mL
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18004254 10 mL 10mL
18034656 25 mL 25mL
18091414 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18004254 Supplier Novus Biologicals™ Supplier No. ACO0010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Affinity resin is designed for simple, rapid His-tagged recombinant protein purification from a cell lysate under native or denaturing conditions

  • Metal chelate affinity chromatography is a rapid one-step purification, which removes most contaminants and can achieve purities close to homogeneity.
  • The rapid purification protocols provided in this handbook along with the NTA resin for affinity chromatography permit the recovery of high levels of pure recombinant protein in minutes.
  • Large numbers of samples can be processed at the same time.
  • This product is manufactured by Expedeon Inc. and distributed by Novus Biologicals.
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Quantity 10 mL
Storage Requirements Store at 4°C. Do not freeze.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.