missing translation for 'onlineSavingsMsg'
Learn More

Aminothiazole, MedChemExpress

Product Code. 16497857
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497857 5 g 5g
16475138 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16497857 Supplier Medchem Express Supplier No. HY12396
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Aminothiazole (2-Aminothiazole), a typical heterocyclic amine, is a precursor for the synthesis of biologically active molecules including sulfur agents, biocides, fungicides, antibiotics, dyes and chemical reaction accelerators.

Aminothiazole (2-Aminothiazole), a typical heterocyclic amine, is a precursor for the synthesis of biologically active molecules including sulfur agents, biocides, fungicides, antibiotics, dyes and chemical reaction accelerators.

Specifications

Chemical Name or Material Aminothiazole
Physical Form Solid
CAS 96-50-4
Color Earth Yellow
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C3H4N2S
For Use With (Application) COVID-19-anti-virus
Synonym 2-Aminothiazole 2-Thiazolylamine
Solubility Information DMSO : ≥ 50 mg/mL (499.30 mM)
SMILES NC1=NC=CS1
Molecular Weight (g/mol) 100.14
Formula Weight 100.14
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.