missing translation for 'onlineSavingsMsg'
Learn More

Aminophenethylamine, TRC

Product Code. 30326807
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326807 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326807 Supplier Toronto Research Chemicals Supplier No. TRCA6139805G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aminophenethylamine
CAS 13472-00-9
Molecular Formula C8H12N2
Quantity 5 g
Synonym 4-Aminobenzeneethanamine, (p-Aminophenyl)ethylamine, 2-(4-Aminophenyl)ethylamine, 2-(4-Aminophenyl)phenethylamine, 2-(p-Aminophenyl)ethylamine4-(2-Aminoethyl)aniline, 4-(2-Aminoethyl)benzenamine, 4-(2-Aminoethyl)phenylamine, 4-Amino-beta-phenethylamine, 4-Amino-beta-phenylethylamine, 4-Aminophenethylamine, 4-Aminophenylethylamine, NSC 299558, NSC 9060, p-(2-Aminoethyl)aniline, p-Aminophenethylamine, beta-(4-Aminophenyl)ethylamine, beta-(p-Aminophenyl)ethylamine
InChI Formula InChI=1S/C8H12N2/c9-6-5-7-1-3-8(10)4-2-7/h1-4H,5-6,9-10H2
SMILES NCCc1ccc(N)cc1
IUPAC Name 4-(2-aminoethyl)aniline
Molecular Weight (g/mol) 136.19
Formula Weight 136.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.