missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminopeptidase PILS/ARTS1 Rabbit anti-Human, Mouse, Rat, Clone: S06-1H3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-14916-100UL
This item is not returnable.
View return policy
Description
Aminopeptidase PILS/ARTS1 Monoclonal antibody specifically detects Aminopeptidase PILS/ARTS1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Specifications
| Aminopeptidase PILS/ARTS1 | |
| Monoclonal | |
| Unconjugated | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| Rabbit | |
| Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| S06-1H3 | |
| Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:20-1:50, Immunohistochemistry-Paraffin, Immunofluorescence | |
| Adipocyte-derived leucine aminopeptidase, A-LAPALAP, Aminopeptidase PILS, ARTS1aminopeptidase regulator of TNFR1 shedding, ARTS-1PILSAP, EC 3.4.11, EC 3.4.11.-, EC 3.4.11.1, endoplasmic reticulum aminopeptidase 1, endoplasmic reticulum aminopeptidase associated with antigen processing, ERAAP1, KIAA0525APPILS, PILS-APERAAP, Puromycin-insensitive leucyl-specific aminopeptidase, Type 1 tumor necrosis factor receptor shedding aminopeptidase regulator | |
| A synthetic peptide of human Aminopeptidase PILS/ARTS1 (Uniprot#: Q9NZ08) | |
| 100 μg | |
| Angiogenesis, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Immunology | |
| 51752 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction