missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Aminopeptidase P2/XPNPEP2 Antibody Pair [HRP]

Product Code. p-7183392 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18287686

Brand: Novus Biologicals™ NBP27937615PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Aminopeptidase P2/XPNPEP2.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 11 pg/mL
Assay Sensitivity 10.94-700 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Aminopeptidase P2/XPNPEP2.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias Aminoacylproline aminopeptidase, APP2, EC 3.4.11.9, mAmP, Membrane-bound aminopeptidase P, Membrane-bound AmP, Membrane-bound APP, xaa-Pro aminopeptidase 2, X-Pro aminopeptidase 2, X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound
Gene ID (Entrez) 7512
Gene Symbol XPNPEP2
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 20 ng of recombinant Aminopeptidase P2/XPNPEP2.
  • Capture Antibody: 1.0 mg/mL of rabbit anti-Aminopeptidase P2/XPNPEP2 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.5 mg/mL rabbit anti-Aminopeptidase P2/XPNPEP2 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Quantity 15 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target Aminopeptidase P2/XPNPEP2
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.7 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.