missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminopeptidase P2/XPNPEP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£222.00 - £470.00
Specifications
| Antigen | Aminopeptidase P2/XPNPEP2 |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18473311
|
Novus Biologicals
NBP1-86535-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18263666
|
Novus Biologicals
NBP1-86535 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aminopeptidase P2/XPNPEP2 Polyclonal specifically detects Aminopeptidase P2/XPNPEP2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Aminopeptidase P2/XPNPEP2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Aminoacylproline aminopeptidase, APP2, EC 3.4.11.9, mAmP, Membrane-bound aminopeptidase P, Membrane-bound AmP, Membrane-bound APP, xaa-Pro aminopeptidase 2, X-Pro aminopeptidase 2, X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound | |
| XPNPEP2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 7512 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VRSQMQKHQKVPTAVLLSALEETAWLFNLRASDIPYNPFFYSYTLLTDSSIRLFANKSRFSSETLSYLNSSCTGPMCVQIEDYSQVRDSIQAYSLGDVRIWIGTSYTMYGI | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title