missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminopeptidase P2/XPNPEP2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99900CL1
This item is not returnable.
View return policy
Description
Aminopeptidase P2/XPNPEP2 Polyclonal antibody specifically detects Aminopeptidase P2/XPNPEP2 in Human samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
Specifications
| Aminopeptidase P2/XPNPEP2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 7512 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| Aminoacylproline aminopeptidase, APP2, EC 3.4.11.9, mAmP, Membrane-bound aminopeptidase P, Membrane-bound AmP, Membrane-bound APP, xaa-Pro aminopeptidase 2, X-Pro aminopeptidase 2, X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound | |
| Produced in rabbits immunized with purified, recombinant Human Aminopeptidase P2/XPNPEP2 (Accession#: O43895; Met 1-Ala 650) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction