missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminopeptidase P1/XPNPEP1 Antibody (SAIC-07B-14) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
£190.00 - £494.00
Specifications
| Antigen | Aminopeptidase P1/XPNPEP1 |
|---|---|
| Clone | SAIC-07B-14 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Mass Spectrometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611555
|
Novus Biologicals
NBP3-20090-0.025mg |
0.025 mg |
£190.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18644954
|
Novus Biologicals
NBP3-20090-0.2mg |
0.2 mg |
£494.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aminopeptidase P1/XPNPEP1 Monoclonal antibody specifically detects Aminopeptidase P1/XPNPEP1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Mass SpectrometrySpecifications
| Aminopeptidase P1/XPNPEP1 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Aminoacylproline aminopeptidase, APP1, cytosolic, Cytosolic aminopeptidase P, EC 3.4.11.9, sAmp, xaa-Pro aminopeptidase 1, XPNPEPLX-prolyl aminopeptidase (aminopeptidase P)-like, X-prolyl aminopeptidase (aminopeptidase P) 1, soluble, X-prolyl aminopeptidase 1, soluble | |
| Peptide 'GSLTFEPLTLVPIQTK' derived from Xaa-Pro aminopeptidase 1 conjugated to KLH. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| SAIC-07B-14 | |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Mass Spectrometry | |
| Unconjugated | |
| Rabbit | |
| metabolism, Signal Transduction | |
| PBS | |
| 7511 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title