missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminopeptidase LRAP/ERAP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-37872
This item is not returnable.
View return policy
Description
Aminopeptidase LRAP/ERAP2 Polyclonal specifically detects Aminopeptidase LRAP/ERAP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Aminopeptidase LRAP/ERAP2 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Q6P179 | |
| ERAP2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RVGLIHDVFQLVGAGRLTLDKALDMTYYLQHETSSPALLEGLSYLESFYHMMDRRNISDISENLKRYLLQYFKPVID | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 3.4.11, EC 3.4.11.3, endoplasmic reticulum aminopeptidase 2, FLJ23633, FLJ23701, FLJ23807, Leukocyte-derived arginine aminopeptidase, LRAPL-RAPEC 3.4.11.- | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 64167 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction