missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminopeptidase LRAP/ERAP2 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81783AF647
This item is not returnable.
View return policy
Description
Aminopeptidase LRAP/ERAP2 Polyclonal antibody specifically detects Aminopeptidase LRAP/ERAP2 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Aminopeptidase LRAP/ERAP2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| Alexa Fluor 647 | |
| EC 3.4.11, EC 3.4.11.3, endoplasmic reticulum aminopeptidase 2, FLJ23633, FLJ23701, FLJ23807, Leukocyte-derived arginine aminopeptidase, LRAPL-RAPEC 3.4.11.- | |
| ERAP2 antibody was raised against a 17 amino acid peptide near the amino terminus of human ERAP2. The immunogen is located within amino acids 110 - 160 of ERAP2 . Amino Acid Squence: LQSEEDSRYMKPGKELK | |
| 0.1 mL | |
| Innate Immunity | |
| 64167 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction