missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminopeptidase A/ENPEP Antibody (6C3), Alexa Fluor™ 700, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Aminopeptidase A/ENPEP Monoclonal antibody specifically detects Aminopeptidase A/ENPEP in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Aminopeptidase A/ENPEP |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 6C3 |
| Conjugate | Alexa Fluor 700 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Aminopeptidase A, APA, AP-A, CD249, CD249 antigen, Differentiation antigen gp160, EAP, EC 3.4.11, EC 3.4.11.7, glutamyl aminopeptidase, glutamyl aminopeptidase (aminopeptidase A), gp160 |
| Host Species | Rat |
| Immunogen | The immunogen for this antibody was Aminopeptidase A/APA. |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?