missing translation for 'onlineSavingsMsg'
Learn More

Aminooxyacetic acid hemihydrochloride, MedChemExpress

Product Code. 16441099
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16441099 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16441099 Supplier Medchem Express Supplier No. HY107994
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Aminooxyacetic acid (Carboxymethoxylamine) hemihydrochloride is a malate-aspartate shuttle (MAS) inhibitor which also inhibits the GABA degradating enzyme GABA-T.

Aminooxyacetic acid (Carboxymethoxylamine) hemihydrochloride is a malate-aspartate shuttle (MAS) inhibitor which also inhibits the GABA degradating enzyme GABA-T.

Specifications

Chemical Name or Material Aminooxyacetic acid hemihydrochloride
Physical Form Crystals
CAS 2921-14-4
Color Off-White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C2H5NO3.1/2HCl
For Use With (Application) Cancer-programmed cell death
Synonym Carboxymethoxylamine hemihydrochlorideAminooxyacetate hemihydrochloride
Solubility Information H2O : 100 mg/mL (914.91 mM; Need ultrasonic) ∣DMSO : 42.9 mg/mL (392.50 mM; Need ultrasonic and warming)
SMILES OC(CON)=O.[H]Cl.[1/2]
Molecular Weight (g/mol) 109.3
Formula Weight 109.3
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.