missing translation for 'onlineSavingsMsg'
Learn More

(Aminooxy)acetic acid hemihydrochloride, MP Biomedicals™

Product Code. 11456492 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456492 5 g 5g
11446492 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456492 Supplier MP Biomedicals Supplier No. 0215010405
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2921-14-4
Molecular Formula C4H11ClN2O6
Molecular Weight (g/mol) 218.59
MDL Number MFCD00012955
InChI Key AXIKLJPWEGNZIG-UHFFFAOYSA-N
Synonym o-carboxymethyl hydroxylamine hemihydrochloride, carboxymethoxylamine hemihydrochloride, aminooxyacetic acid hemihydrochloride, 2-aminooxy acetic acid hydrochloride 2:1, unii-5g5qi2gn7a, acetic acid, aminooxy-, hydrochloride 2:1, aminooxy acetic acid hemihydrochloride, 5g5qi2gn7a, carboxymethoxyamine hemihydrochloride
PubChem CID 2723609
IUPAC Name 2-(aminooxy)acetic acid 2-(azaniumyloxy)acetic acid chloride
SMILES [Cl-].NOCC(O)=O.[NH3+]OCC(O)=O

Specifications

Color White
Assay Percent Range 99%
Quantity 5 g
Formula Weight 109.3
Physical Form Crystalline Powder
Chemical Name or Material (Aminooxy)Acetic Acid Hemihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.