missing translation for 'onlineSavingsMsg'
Learn More

(Aminomethyl)cyclopropane, 97%, Thermo Scientific Chemicals

Product Code. 10666271
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2516-47-4
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.11
InChI Key IGSKHXTUVXSOMB-UHFFFAOYSA-N
Synonym cyclopropylmethylamine, cyclopropanemethylamine, cyclopropanemethanamine, aminomethylcyclopropane, aminomethyl cyclopropane, 1-cyclopropylmethanamine, cyclopropane methyl amine, cyclopropylmethylamin, cyclopropanmethylamine
PubChem CID 75646
IUPAC Name cyclopropylmethanamine
SMILES C1CC1CN

Specifications

Color Colorless to Yellow
Density 0.8200g/mL
Boiling Point 84.0°C to 85.0°C
Flash Point −30°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4320 to 1.4340
Quantity 10 g
Specific Gravity 0.82
Formula Weight 71.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (Aminomethyl)cyclopropane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Aminomethylcyclopropane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'