missing translation for 'onlineSavingsMsg'
Learn More

Aminomethylalizarindiacetic Acid Reagent (Sol A+B+C) 50 mL+ 50 mL + 100 mL - British Pharmacopoeia, Fisher Chemical™

Product Code. 30695076 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
200 mL
Unit Size:
200mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30695076 200 mL 200mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30695076 Supplier Fisher Chemical Supplier No. BPR 013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Pharmacopoeia reagents for testing and monographs, produced under a GMP-compliant ISO 9001 quality management system.

    This Pharmacopoeia reagent is manufactured :
  • Using pharmacopoeia reagent quality components
  • Water meeting the relevant pharmacopoeia monograph requirement for Purified Water
  • Supplied with a Certificate of Analysis for Performance Test
  • Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO 17034

Specifications

Quantity 200 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.