Learn More
(Aminomethyl)cyclopropane hydrochloride, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 7252-53-1 |
---|---|
Molecular Formula | C4H9ClN |
Molecular Weight (g/mol) | 106.57 |
MDL Number | MFCD00012544 |
InChI Key | MQCZBGGAZQXTLH-UHFFFAOYSA-N |
Synonym | cyclopropanemethylamine hydrochloride, cyclopropylmethanamine hydrochloride, cyclopropylmethyl ammonium chloride, cyclopropylmethylamine hydrochloride, aminomethylcyclopropane hydrochloride, 1-cyclopropylmethanamine hydrochloride, aminomethyl cyclopropane hydrochloride, cyclopropylmethanaminium chloride, cyclopropylmethylamin hydrochloride, cyclopropylmethanamine-hydrochloride |
PubChem CID | 81671 |
SMILES | [Cl].NCC1CC1 |
Description
Chemical Identifiers
7252-53-1 | |
106.57 | |
MQCZBGGAZQXTLH-UHFFFAOYSA-N | |
81671 |
C4H9ClN | |
MFCD00012544 | |
cyclopropanemethylamine hydrochloride, cyclopropylmethanamine hydrochloride, cyclopropylmethyl ammonium chloride, cyclopropylmethylamine hydrochloride, aminomethylcyclopropane hydrochloride, 1-cyclopropylmethanamine hydrochloride, aminomethyl cyclopropane hydrochloride, cyclopropylmethanaminium chloride, cyclopropylmethylamin hydrochloride, cyclopropylmethanamine-hydrochloride | |
[Cl].NCC1CC1 |
Specifications
7252-53-1 | |
100.0 | |
White | |
Authentic | |
C4H9ClN | |
MFCD00012544 | |
cyclopropanemethylamine hydrochloride, cyclopropylmethanamine hydrochloride, cyclopropylmethyl ammonium chloride, cyclopropylmethylamine hydrochloride, aminomethylcyclopropane hydrochloride, 1-cyclopropylmethanamine hydrochloride, aminomethyl cyclopropane hydrochloride, cyclopropylmethanaminium chloride, cyclopropylmethylamin hydrochloride, cyclopropylmethanamine-hydrochloride | |
[Cl].NCC1CC1 | |
106.57 | |
107.58 | |
Powder |
96.0 | |
205.0°C to 209.0°C | |
42.87 to 46.44% C, 12.50 to 13.54% N | |
97% | |
C3H5CH2NH2·HCl | |
12,4 | |
MQCZBGGAZQXTLH-UHFFFAOYSA-N | |
1-cyclopropylmethanamine hydrochloridyl | |
81671 | |
97% | |
(Aminomethyl)cyclopropane hydrochloride |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 230-668-
- (Aminomethyl)cyclopropane hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only