missing translation for 'onlineSavingsMsg'
Learn More

(Aminomethyl)cyclopropane, 97%

Product Code. 10622983 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10622983 5 g Glass bottle 5g
10666271 10 g Glass bottle 10g
11915201 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10622983 Supplier Thermo Scientific Acros Supplier No. 258720050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2516-47-4 | C4H9N | 71.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2516-47-4
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.11
InChI Key IGSKHXTUVXSOMB-UHFFFAOYSA-N
Synonym cyclopropylmethylamine, cyclopropanemethylamine, cyclopropanemethanamine, aminomethylcyclopropane, aminomethyl cyclopropane, 1-cyclopropylmethanamine, cyclopropane methyl amine, cyclopropylmethylamin, cyclopropanmethylamine
PubChem CID 75646
IUPAC Name cyclopropylmethanamine
SMILES C1CC1CN

Specifications

Color Colorless to Yellow
Density 0.8200g/mL
Boiling Point 84.0°C to 85.0°C
Flash Point −30°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4320 to 1.4340
Quantity 5 g
Specific Gravity 0.82
Formula Weight 71.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (Aminomethyl)cyclopropane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.