missing translation for 'onlineSavingsMsg'
Learn More

Aminolevulinic Acid Related Compound B, TRC

Product Code. 30138056
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138056 1 g 1g
30136481 250 mg 250mg
30003838 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138056 Supplier Toronto Research Chemicals Supplier No. TRCA6408331G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109258-71-1
Molecular Formula C14H13NO5
Molecular Weight (g/mol) 275
Synonym Methyl 5-(1,3-dioxoisoindolin-2-yl)-4-oxopentanoate
IUPAC Name methyl 5-(1,3-dioxoisoindol-2-yl)-4-oxopentanoate
SMILES COC(=O)CCC(=O)CN1C(=O)c2ccccc2C1=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H13NO5/c1-20-12(17)7-6-9(16)8-15-13(18)10-4-2-3-5-11(10)14(15)19/h2-5H,6-8H2,1H3
Formula Weight 275.0794
Chemical Name or Material Aminolevulinic Acid Related Compound B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.